The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025260.3 | NP_079536.2 | 5 - 6 | 684 | 85 |
NM_138272.2 | NP_612116.1 | 5 - 6 | 665 | 85 | |
NM_138273.2 | NP_612117.1 | 4 - 5 | 593 | 85 | |
NM_138274.2 | NP_612118.1 | 3 - 4 | 552 | 85 | |
NM_138275.2 | NP_612119.1 | 3 - 4 | 533 | 85 | |
XM_011514920.2 | XP_011513222.1 | 5 - 6 | 699 | 85 | |
XM_011514921.2 | XP_011513223.1 | 5 - 6 | 718 | 85 | |
XM_017011330.1 | XP_016866819.1 | 3 - 4 | 745 | 85 | |
XM_017011331.1 | XP_016866820.1 | 3 - 4 | 983 | 85 | |
XM_017011332.1 | XP_016866821.1 | 4 - 5 | 627 | 85 | |
GenBank mRNA | AJ292259.1 | - | 5 - 6 | 644 | 85 |
AJ292260.1 | - | 5 - 6 | 625 | 85 | |
AJ292261.1 | - | 4 - 5 | 553 | 85 | |
AJ292262.1 | - | 3 - 4 | 512 | 85 | |
AJ292263.1 | - | 3 - 4 | 493 | 85 | |
AJ292264.1 | - | 4 - 5 | 762 | 85 | |
AK092392.1 | - | 5 - 6 | 655 | 85 |