The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138273.2 | NP_612117.1 | 2 - 3 | 450 | 112 |
XM_017011332.1 | XP_016866821.1 | 2 - 3 | 484 | 112 | |
GenBank mRNA | AJ292261.1 | - | 2 - 3 | 410 | 112 |