The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004212.3 | NP_004203.2 | 17 - 18 | 1919 | 101 |
XM_011522198.2 | XP_011520500.1 | 17 - 18 | 1939 | 101 | |
XM_011522199.1 | XP_011520501.1 | 16 - 17 | 1791 | 101 | |
XM_011522200.2 | XP_011520502.1 | 15 - 16 | 1709 | 101 | |
XM_011522202.2 | XP_011520504.1 | 9 - 10 | 1090 | 101 | |
GenBank mRNA | AF036109.1 | - | 16 - 17 | 1868 | 101 |
AK025121.1 | - | 1 - 2 | 1006 | 101 | |
AK222709.1 | - | 17 - 18 | 1919 | 101 | |
AK291974.1 | - | 17 - 18 | 1890 | 101 | |
BC093737.1 | - | 16 - 17 | 1882 | 101 | |
BC112047.1 | - | 16 - 17 | 1894 | 101 | |
U84392.1 | - | 17 - 18 | 1913 | 101 |