The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004212.3 | NP_004203.2 | 7 - 8 | 770 | 93 |
XM_011522198.2 | XP_011520500.1 | 7 - 8 | 790 | 93 | |
XM_011522199.1 | XP_011520501.1 | 7 - 8 | 741 | 93 | |
XM_011522200.2 | XP_011520502.1 | 7 - 8 | 741 | 93 | |
XM_011522201.2 | XP_011520503.1 | 7 - 8 | 741 | 93 | |
GenBank mRNA | AF036109.1 | - | 6 - 7 | 719 | 93 |
AK222709.1 | - | 7 - 8 | 770 | 93 | |
AK291974.1 | - | 7 - 8 | 741 | 93 | |
BC093737.1 | - | 6 - 7 | 733 | 93 | |
BC112047.1 | - | 6 - 7 | 745 | 93 | |
U84392.1 | - | 7 - 8 | 764 | 93 |