Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
Chromosome Location
Chr.12: 6638076 - 6647414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032489.2 NP_115878.2 5 - 6 991 112
GenBank mRNA AK302290.1 - 5 - 6 889 112
AK311572.1 - 5 - 6 798 112
AK312782.1 - 5 - 6 1009 112
BC033010.2 - 5 - 6 988 112

View Full Product Details