Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DDX12P
- Entrez Gene ID
-
440081
- Gene Name
- DEAD/H-box helicase 12, pseudogene
- Gene Aliases
-
CHLR2, DDX12
- Chromosome Location
-
Chr.12: 9417691 - 9448172 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033399.1 |
|
26 - 27 |
2904 |
59 |
- Gene Symbol
- DDX11
- Entrez Gene ID
-
1663
- Gene Name
- DEAD/H-box helicase 11
- Gene Aliases
-
CHL1, CHLR1, KRG2, WABS
- Chromosome Location
-
Chr.12: 31073574 - 31104799 on Build GRCh38
View Full Product Details