The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138423.3 | NP_612432.2 | 8 - 9 | 1392 | 84 |
GenBank mRNA | AK298564.1 | - | 9 - 10 | 1107 | 84 |
AL832333.1 | - | 8 - 9 | 1164 | 84 | |
AL832840.1 | - | 8 - 9 | 834 | 84 | |
BC041786.1 | - | 2 - 3 | 270 | 84 | |
BC063480.1 | - | 8 - 9 | 1346 | 84 |