The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138423.3 | NP_612432.2 | 9 - 10 | 1557 | 75 |
GenBank mRNA | AK298564.1 | - | 10 - 11 | 1272 | 75 |
AL832333.1 | - | 9 - 10 | 1329 | 75 | |
AL832840.1 | - | 9 - 10 | 999 | 75 | |
BC041786.1 | - | 3 - 4 | 435 | 75 |