Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASC4
Entrez Gene ID
113201
Gene Name
cancer susceptibility candidate 4
Gene Aliases
H63
Chromosome Location
Chr.15: 44288616 - 44415761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138423.3 NP_612432.2 4 - 5 896 184
NM_177974.2 NP_816929.1 4 - 5 896 184
XM_017021880.1 XP_016877369.1 4 - 5 991 184
GenBank mRNA AK298564.1 - 5 - 6 611 184
AL832333.1 - 4 - 5 668 184
AL832840.1 - 4 - 5 338 184
AY358086.1 - 4 - 5 847 184
BC063480.1 - 4 - 5 850 184

View Full Product Details