Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLEKHG6
- Entrez Gene ID
-
55200
- Gene Name
- pleckstrin homology and RhoGEF domain containing G6
- Gene Aliases
-
MyoGEF
- Chromosome Location
-
Chr.12: 6310436 - 6328506 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF289613.1 |
- |
1 - 2 |
217 |
61 |
View Full Product Details