The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014753.3 | NP_055568.3 | 10 - 11 | 2344 | 144 |
XM_005271846.3 | XP_005271903.1 | 10 - 11 | 2219 | 144 | |
XM_005271848.3 | XP_005271905.1 | 10 - 11 | 2075 | 144 | |
XM_005271849.3 | XP_005271906.1 | 10 - 11 | 2075 | 144 | |
XM_011540402.2 | XP_011538704.1 | 10 - 11 | 2075 | 144 | |
XM_011540403.2 | XP_011538705.1 | 2 - 3 | 878 | 144 | |
GenBank mRNA | AK308420.1 | - | 10 - 11 | 2051 | 144 |
BC043345.1 | - | 10 - 11 | 2039 | 144 | |
BC150252.1 | - | 9 - 10 | 2013 | 144 | |
D80009.1 | - | 9 - 10 | 2013 | 144 |