Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMS1
Entrez Gene ID
9790
Gene Name
BMS1, ribosome biogenesis factor
Gene Aliases
ACC, BMS1L
Chromosome Location
Chr.10: 42782506 - 42834937 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014753.3 NP_055568.3 13 - 14 2689 93
XM_005271846.3 XP_005271903.1 13 - 14 2564 93
XM_005271848.3 XP_005271905.1 13 - 14 2420 93
XM_005271849.3 XP_005271906.1 13 - 14 2420 93
XM_011540402.2 XP_011538704.1 13 - 14 2420 93
XM_011540403.2 XP_011538705.1 5 - 6 1223 93
GenBank mRNA AK308420.1 - 13 - 14 2396 93
BC043345.1 - 13 - 14 2384 93
BC150252.1 - 12 - 13 2358 93
D80009.1 - 12 - 13 2358 93

View Full Product Details