Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF2BP
Entrez Gene ID
11077
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
Chromosome Location
Chr.21: 43453987 - 43659499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007031.1 NP_008962.1 2 - 3 365 168
XM_011529443.2 XP_011527745.2 2 - 3 371 168
XM_011529447.2 XP_011527749.1 2 - 3 371 168
XM_017028267.1 XP_016883756.1 2 - 3 371 168
XM_017028268.1 XP_016883757.1 2 - 3 371 168
XM_017028269.1 XP_016883758.1 2 - 3 371 168
XM_017028270.1 XP_016883759.1 2 - 3 371 168
XM_017028272.1 XP_016883761.1 2 - 3 371 168
GenBank mRNA AB007131.1 - 2 - 3 365 168
AK314121.1 - 2 - 3 360 168
BC000153.2 - 2 - 3 348 168

View Full Product Details