The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256295.1 | NP_001243224.1 | 5 - 6 | 769 | 56 |
NM_005239.5 | NP_005230.1 | 4 - 5 | 599 | 56 | |
XM_005260935.1 | XP_005260992.1 | 4 - 5 | 672 | 56 | |
XM_017028290.1 | XP_016883779.1 | 4 - 5 | 3269 | 56 | |
GenBank mRNA | AK096841.1 | - | 5 - 6 | 769 | 56 |
AK315563.1 | - | 4 - 5 | 502 | 56 | |
BC017040.1 | - | 4 - 5 | 471 | 56 | |
BC042954.2 | - | 4 - 5 | 501 | 56 | |
BT006838.1 | - | 3 - 4 | 309 | 56 | |
J04102.1 | - | 4 - 5 | 600 | 56 |