The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277196.1 | NP_001264125.1 | 14 - 15 | 2148 | 92 |
NM_016018.4 | NP_057102.4 | 15 - 16 | 2226 | 92 | |
XM_005250931.1 | XP_005250988.1 | 15 - 16 | 2211 | 92 | |
XM_011517075.1 | XP_011515377.1 | 15 - 16 | 2211 | 92 | |
XM_011517076.1 | XP_011515378.1 | 15 - 16 | 2207 | 92 | |
XM_011517077.1 | XP_011515379.1 | 15 - 16 | 2208 | 92 | |
XM_011517078.1 | XP_011515380.1 | 15 - 16 | 2199 | 92 | |
XM_011517079.1 | XP_011515381.1 | 15 - 16 | 2196 | 92 | |
XM_011517080.1 | XP_011515382.1 | 14 - 15 | 2121 | 92 | |
XM_011517081.1 | XP_011515383.1 | 14 - 15 | 2118 | 92 | |
XM_017013509.1 | XP_016868998.1 | 15 - 16 | 2192 | 92 | |
XM_017013510.1 | XP_016868999.1 | 14 - 15 | 2117 | 92 | |
XM_017013511.1 | XP_016869000.1 | 14 - 15 | 2114 | 92 | |
XM_017013512.1 | XP_016869001.1 | 14 - 15 | 2114 | 92 | |
XM_017013513.1 | XP_016869002.1 | 15 - 16 | 5034 | 92 | |
XM_017013514.1 | XP_016869003.1 | 15 - 16 | 5030 | 92 | |
XM_017013515.1 | XP_016869004.1 | 8 - 9 | 1665 | 92 | |
GenBank mRNA | AK057961.1 | - | 2 - 3 | 240 | 92 |
AY279109.1 | - | 2 - 3 | 240 | 92 | |
BC050655.1 | - | 1 - 2 | 65 | 92 | |
DA196349.1 | - | 3 - 4 | 300 | 92 |