Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20L1
Entrez Gene ID
51105
Gene Name
PHD finger protein 20-like 1
Gene Aliases
CGI-72, TDRD20B, URLC1
Chromosome Location
Chr.8: 132775358 - 132848807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277196.1 NP_001264125.1 4 - 5 658 110
NM_016018.4 NP_057102.4 4 - 5 658 110
NM_198513.1 NP_940915.1 4 - 5 654 110
XM_005250931.1 XP_005250988.1 4 - 5 628 110
XM_011517075.1 XP_011515377.1 4 - 5 628 110
XM_011517076.1 XP_011515378.1 4 - 5 624 110
XM_011517077.1 XP_011515379.1 4 - 5 628 110
XM_011517078.1 XP_011515380.1 4 - 5 628 110
XM_011517079.1 XP_011515381.1 4 - 5 628 110
XM_011517080.1 XP_011515382.1 4 - 5 628 110
XM_011517081.1 XP_011515383.1 4 - 5 628 110
XM_011517088.1 XP_011515390.1 4 - 5 628 110
XM_017013509.1 XP_016868998.1 4 - 5 624 110
XM_017013510.1 XP_016868999.1 4 - 5 624 110
XM_017013511.1 XP_016869000.1 4 - 5 624 110
XM_017013512.1 XP_016869001.1 4 - 5 624 110
XM_017013516.1 XP_016869005.1 4 - 5 628 110
XM_017013517.1 XP_016869006.1 4 - 5 628 110
GenBank mRNA AB101203.1 - 4 - 5 625 110
AK022280.1 - 3 - 4 339 110
AK290506.1 - 4 - 5 628 110
AL538135.3 - 4 - 5 614 110
BC002822.2 - 4 - 5 616 110
BC015211.1 - 4 - 5 604 110

View Full Product Details