The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006446.4 | NP_006437.3 | 8 - 9 | 1075 | 152 |
GenBank mRNA | AB026257.1 | - | 8 - 9 | 1070 | 152 |
AF060500.1 | - | 7 - 8 | 971 | 152 | |
AF205071.1 | - | 8 - 9 | 1105 | 152 | |
AJ132573.1 | - | 7 - 8 | 1034 | 152 | |
AK312970.1 | - | 8 - 9 | 1057 | 152 | |
BC114376.1 | - | 8 - 9 | 1070 | 152 |