The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030762.2 | NP_110389.1 | 2 - 3 | 418 | 113 |
GenBank mRNA | AB044088.1 | - | 2 - 3 | 261 | 113 |
AK303677.1 | - | 2 - 3 | 257 | 113 | |
AY665467.1 | - | 2 - 3 | 228 | 113 | |
BC025968.1 | - | 2 - 3 | 418 | 113 |