The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002907.3 | NP_002898.2 | 1 - 2 | 499 | 100 |
NM_032941.2 | NP_116559.1 | 2 - 3 | 302 | 100 | |
XM_005253461.3 | XP_005253518.1 | 2 - 3 | 373 | 100 | |
XM_005253462.4 | XP_005253519.1 | 2 - 3 | 188 | 100 | |
GenBank mRNA | AK291627.1 | - | 1 - 2 | 423 | 100 |
AV718094.1 | - | 2 - 3 | 234 | 100 | |
BC001052.2 | - | 2 - 3 | 184 | 100 | |
BE794392.1 | - | 2 - 3 | 165 | 100 | |
CN414472.1 | - | 1 - 2 | 440 | 100 | |
DA028052.1 | - | 1 - 2 | 465 | 100 | |
DA547846.1 | - | 2 - 3 | 409 | 100 |