The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002907.3 | NP_002898.2 | 4 - 5 | 933 | 76 |
NM_032941.2 | NP_116559.1 | 5 - 6 | 736 | 76 | |
XM_005253461.3 | XP_005253518.1 | 5 - 6 | 807 | 76 | |
XM_005253462.4 | XP_005253519.1 | 5 - 6 | 622 | 76 | |
XM_005253463.3 | XP_005253520.1 | 4 - 5 | 656 | 76 | |
XM_005253464.3 | XP_005253521.1 | 4 - 5 | 606 | 76 | |
GenBank mRNA | AK291627.1 | - | 4 - 5 | 857 | 76 |
AY157499.1 | - | 4 - 5 | 512 | 76 | |
BC001052.2 | - | 5 - 6 | 618 | 76 | |
BT007119.1 | - | 3 - 4 | 388 | 76 | |
D37984.1 | - | 2 - 3 | 388 | 76 | |
L36140.1 | - | 4 - 5 | 915 | 76 |