Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAC2
Entrez Gene ID
5880
Gene Name
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Gene Aliases
EN-7, Gx, HSPC022, p21-Rac2
Chromosome Location
Chr.22: 37225261 - 37244299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002872.4 NP_002863.1 7 - 7 877 131
GenBank mRNA AF077208.1 - 1 - 1 100 131
BC001485.2 - 7 - 7 849 131
BC018735.1 - 3 - 3 396 131
BC051899.1 - 1 - 1 117 131

View Full Product Details