The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190458.1 | NP_001177387.1 | 35 - 36 | 4591 | 140 |
NM_001193536.1 | NP_001180465.1 | 36 - 37 | 4687 | 140 | |
NM_203447.3 | NP_982272.2 | 37 - 38 | 4902 | 140 | |
XM_011518045.2 | XP_011516347.1 | 36 - 37 | 4661 | 140 | |
XM_011518046.2 | XP_011516348.1 | 37 - 38 | 4800 | 140 | |
XM_011518047.2 | XP_011516349.1 | 37 - 38 | 4983 | 140 | |
XM_011518048.2 | XP_011516350.1 | 36 - 37 | 4722 | 140 | |
XM_011518049.2 | XP_011516351.1 | 24 - 25 | 3462 | 140 | |
XM_017015173.1 | XP_016870662.1 | 37 - 38 | 4757 | 140 | |
XM_017015174.1 | XP_016870663.1 | 37 - 38 | 4837 | 140 | |
GenBank mRNA | AB191037.1 | - | 37 - 38 | 4902 | 140 |
AK024436.1 | - | 16 - 17 | 2034 | 140 | |
AK074081.1 | - | 23 - 24 | 3020 | 140 | |
AK090429.1 | - | 29 - 30 | 3891 | 140 | |
AL832270.1 | - | 35 - 36 | 6743 | 140 | |
AY117695.1 | - | 24 - 25 | 3578 | 140 | |
BC019102.2 | - | 2 - 3 | 311 | 140 | |
BC112894.1 | - | 6 - 7 | 767 | 140 | |
BC130518.1 | - | 36 - 37 | 4569 | 140 | |
BC143929.1 | - | 36 - 37 | 4569 | 140 |