Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK8
Entrez Gene ID
81704
Gene Name
dedicator of cytokinesis 8
Gene Aliases
HEL-205, MRD2, ZIR8
Chromosome Location
Chr.9: 211297 - 465260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190458.1 NP_001177387.1 7 - 8 793 68
NM_001193536.1 NP_001180465.1 7 - 8 793 68
NM_203447.3 NP_982272.2 8 - 9 1008 68
XM_011518045.2 XP_011516347.1 8 - 9 863 68
XM_011518046.2 XP_011516348.1 8 - 9 906 68
XM_011518047.2 XP_011516349.1 8 - 9 1089 68
XM_011518048.2 XP_011516350.1 7 - 8 828 68
XM_017015173.1 XP_016870662.1 8 - 9 863 68
XM_017015174.1 XP_016870663.1 8 - 9 943 68
GenBank mRNA AB191037.1 - 8 - 9 1008 68
AK093572.1 - 7 - 8 791 68
AK293353.1 - 8 - 9 1005 68
AL832270.1 - 7 - 8 757 68
BC045629.1 - 7 - 8 782 68
BC130518.1 - 7 - 8 771 68
BC143929.1 - 7 - 8 771 68
HM991480.1 - 7 - 8 789 68
KC736820.1 - 6 - 7 692 68

View Full Product Details