The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002156.4 | NP_002147.2 | 5 - 6 | 738 | 140 |
NM_199440.1 | NP_955472.1 | 5 - 6 | 718 | 140 | |
GenBank mRNA | AK297305.1 | - | - | 2191 | 140 |
AK298368.1 | - | 5 - 6 | 458 | 140 | |
AK298632.1 | - | 5 - 6 | 661 | 140 | |
AK298844.1 | - | 4 - 5 | 489 | 140 | |
BC002676.2 | - | 5 - 6 | 648 | 140 | |
BC003030.1 | - | 5 - 6 | 647 | 140 | |
BC047350.1 | - | 4 - 5 | 808 | 140 | |
BC067082.1 | - | 5 - 6 | 646 | 140 | |
BC073746.1 | - | 5 - 6 | 646 | 140 | |
BX647145.1 | - | 4 - 5 | 1403 | 140 | |
EU719626.1 | - | 4 - 5 | 601 | 140 | |
KU178139.1 | - | 4 - 5 | 601 | 140 | |
M22382.1 | - | 5 - 6 | 646 | 140 | |
M34664.1 | - | 5 - 6 | 625 | 140 |