The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243313.1 | NP_001230242.1 | 23 - 24 | 2982 | 100 |
NM_015690.4 | NP_056505.2 | 23 - 24 | 3045 | 100 | |
XM_005246464.1 | XP_005246521.1 | 23 - 24 | 2933 | 100 | |
XM_011510959.1 | XP_011509261.1 | 23 - 24 | 3018 | 100 | |
XM_011510960.1 | XP_011509262.1 | 23 - 24 | 2986 | 100 | |
XM_011510961.1 | XP_011509263.1 | 14 - 15 | 1616 | 100 | |
XM_017003804.1 | XP_016859293.1 | 23 - 24 | 2870 | 100 | |
XM_017003805.1 | XP_016859294.1 | 22 - 23 | 2722 | 100 | |
GenBank mRNA | AB033104.1 | - | 23 - 24 | 2939 | 100 |
AF200815.1 | - | 22 - 23 | 2766 | 100 | |
AK022692.1 | - | 2 - 3 | 254 | 100 | |
AL133630.1 | - | 3 - 4 | 301 | 100 | |
BC026158.2 | - | 23 - 24 | 2985 | 100 |