Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STK36
Entrez Gene ID
27148
Gene Name
serine/threonine kinase 36
Gene Aliases
FU
Chromosome Location
Chr.2: 218672026 - 218702717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK022692.1 - 3 - 4 397 61

View Full Product Details