Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP58
Entrez Gene ID
51602
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
HSPC120, NOP5, NOP5/NOP58
Chromosome Location
Chr.2: 202265716 - 202303661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015934.4 NP_057018.1 8 - 9 1006 88
GenBank mRNA AF123534.1 - 8 - 9 780 88
AF263608.1 - 8 - 9 780 88
AK023975.1 - 7 - 8 705 88
AK307396.1 - 8 - 9 986 88
AL117554.1 - 8 - 9 989 88
BC001707.1 - 8 - 9 972 88
BC009306.2 - 8 - 9 936 88
BC018805.1 - 8 - 9 959 88
BC032592.2 - 8 - 9 930 88

View Full Product Details