Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173431.1 NP_001166902.1 13 - 14 4656 68
NM_015311.2 NP_056126.1 13 - 14 4656 68
XM_005246424.4 XP_005246481.1 12 - 13 4404 68
XM_005246427.4 XP_005246484.1 11 - 12 4119 68
XM_011510857.2 XP_011509159.1 13 - 14 4683 68
XM_011510863.2 XP_011509165.1 13 - 14 4675 68
XM_017003696.1 XP_016859185.1 13 - 14 4682 68
XM_017003697.1 XP_016859186.1 13 - 14 4682 68
XM_017003698.1 XP_016859187.1 12 - 13 4405 68
XM_017003699.1 XP_016859188.1 12 - 13 4406 68
XM_017003700.1 XP_016859189.1 11 - 12 4128 68
GenBank mRNA AK023854.1 - 4 - 5 1047 68
AK299261.1 - 4 - 5 948 68
EF063637.1 - 13 - 14 4402 68
EF063638.1 - 13 - 14 4399 68

View Full Product Details