The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195731.1 | NP_001182660.1 | 2 - 3 | 760 | 81 |
NM_024536.5 | NP_078812.2 | 2 - 3 | 1116 | 81 | |
GenBank mRNA | AB086063.1 | - | 2 - 3 | 950 | 81 |
AB095813.1 | - | 2 - 3 | 1125 | 81 | |
AK302124.1 | - | 2 - 3 | 760 | 81 | |
AL136814.1 | - | 1 - 2 | 263 | 81 | |
AY358403.1 | - | 2 - 3 | 1116 | 81 | |
BC021223.2 | - | 2 - 3 | 1116 | 81 | |
BC023531.2 | - | 2 - 3 | 1067 | 81 |