The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173408.1 | NP_001166879.1 | 3 - 4 | 1853 | 89 |
NM_001173431.1 | NP_001166902.1 | 3 - 4 | 1853 | 89 | |
NM_015311.2 | NP_056126.1 | 3 - 4 | 1853 | 89 | |
XM_005246424.4 | XP_005246481.1 | 3 - 4 | 1877 | 89 | |
XM_005246427.4 | XP_005246484.1 | 3 - 4 | 1868 | 89 | |
XM_011510857.2 | XP_011509159.1 | 3 - 4 | 1880 | 89 | |
XM_011510863.2 | XP_011509165.1 | 3 - 4 | 1872 | 89 | |
XM_011510864.2 | XP_011509166.1 | 3 - 4 | 1872 | 89 | |
XM_011510865.2 | XP_011509167.1 | 3 - 4 | 1872 | 89 | |
XM_011510866.2 | XP_011509168.1 | 3 - 4 | 1871 | 89 | |
XM_017003696.1 | XP_016859185.1 | 3 - 4 | 1879 | 89 | |
XM_017003697.1 | XP_016859186.1 | 3 - 4 | 1879 | 89 | |
XM_017003698.1 | XP_016859187.1 | 3 - 4 | 1878 | 89 | |
XM_017003699.1 | XP_016859188.1 | 3 - 4 | 1879 | 89 | |
XM_017003700.1 | XP_016859189.1 | 3 - 4 | 1877 | 89 | |
GenBank mRNA | AB014557.2 | - | 3 - 4 | 1048 | 89 |
AK025946.1 | - | 2 - 3 | 306 | 89 | |
BC007201.1 | - | 3 - 4 | 1598 | 89 | |
EF063638.1 | - | 3 - 4 | 1596 | 89 |