Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173431.1 NP_001166902.1 12 - 13 4383 84
NM_015311.2 NP_056126.1 12 - 13 4383 84
XM_011510857.2 XP_011509159.1 12 - 13 4410 84
XM_011510863.2 XP_011509165.1 12 - 13 4402 84
XM_011510864.2 XP_011509166.1 12 - 13 4402 84
XM_011510865.2 XP_011509167.1 12 - 13 4402 84
XM_011510866.2 XP_011509168.1 11 - 12 4125 84
XM_017003696.1 XP_016859185.1 12 - 13 4409 84
XM_017003697.1 XP_016859186.1 12 - 13 4409 84
XM_017003699.1 XP_016859188.1 11 - 12 4133 84
GenBank mRNA AB014557.2 - 11 - 12 3302 84
EF063637.1 - 12 - 13 4129 84
EF063638.1 - 12 - 13 4126 84

View Full Product Details