Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011510864.2 XP_011509166.1 13 - 14 4544 88
XM_011510865.2 XP_011509167.1 13 - 14 4544 88
XM_011510866.2 XP_011509168.1 12 - 13 4267 88
GenBank mRNA AB014557.2 - 12 - 13 3444 88

View Full Product Details