The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077198.2 | NP_001070666.1 | 9 - 10 | 1654 | 91 |
NM_024085.4 | NP_076990.4 | 8 - 9 | 1602 | 91 | |
GenBank mRNA | AK025822.1 | - | 3 - 4 | 956 | 91 |
AK056595.1 | - | 9 - 10 | 1595 | 91 | |
AK289755.1 | - | 9 - 10 | 1549 | 91 | |
AK302521.1 | - | 8 - 9 | 1493 | 91 | |
AK315553.1 | - | 3 - 4 | 966 | 91 | |
AL833865.1 | - | 2 - 3 | 851 | 91 | |
BC001098.2 | - | 1 - 2 | 129 | 91 | |
BX537984.1 | - | 8 - 9 | 1523 | 91 | |
BX538192.1 | - | 8 - 9 | 1497 | 91 | |
BX538198.1 | - | 9 - 10 | 1431 | 91 |