The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040020.1 | NP_001035109.1 | 8 - 9 | 668 | 111 |
NM_014888.2 | NP_055703.1 | 8 - 9 | 708 | 111 | |
XM_011515736.2 | XP_011514038.1 | 9 - 10 | 769 | 111 | |
XM_011515737.2 | XP_011514039.1 | 9 - 10 | 800 | 111 | |
GenBank mRNA | AK290682.1 | - | 8 - 9 | 663 | 111 |
AK292731.1 | - | 8 - 9 | 648 | 111 | |
BC024200.1 | - | 8 - 9 | 641 | 111 | |
BC046932.1 | - | 8 - 9 | 636 | 111 | |
BC068526.1 | - | 8 - 9 | 674 | 111 | |
D87120.1 | - | 1 - 1 | 627 | 111 |