Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
Chromosome Location
Chr.2: 101991805 - 102028544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011511804.2 XP_011510106.1 1 - 2 220 77

View Full Product Details