Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK021642.1 - 1 464 Chr. 12 - 82704039 - 82706323 [+] on Build GRCh38 131
GenBankmRNA AU118438.1 - 1 464 Chr. 12 - 82704039 - 82704833 [+] on Build GRCh38 131


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transmembrane and tetratricopeptide repeat containing 2 TMTC2 IBDBP1 Sense Chr. - 82686952 - 83134868 on Build GRCh38
Downstream solute carrier family 6 member 15 SLC6A15 NTT73, SBAT1, V7-3, hv7-3 2.2mb Anti-sense Chr. - 84859488 - 84912876 on Build GRCh38

View Full Product Details