Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP9
Entrez Gene ID
64333
Gene Name
Rho GTPase activating protein 9
Gene Aliases
10C, RGL1
Chromosome Location
Chr.12: 57472255 - 57488814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080156.2 NP_001073625.1 6 - 7 995 115
NM_001080157.1 NP_001073626.1 7 - 8 1350 115
NM_001319850.1 NP_001306779.1 11 - 12 1521 115
NM_001319851.1 NP_001306780.1 5 - 6 1214 115
NM_001319852.1 NP_001306781.1 6 - 7 995 115
NM_032496.3 NP_115885.2 8 - 9 1276 115
XM_005269083.2 XP_005269140.1 6 - 7 660 115
XM_005269084.2 XP_005269141.1 5 - 6 593 115
XM_005269085.2 XP_005269142.1 5 - 6 567 115
XM_011538656.2 XP_011536958.1 10 - 11 1313 115
XM_011538657.2 XP_011536959.1 7 - 8 1097 115
XM_011538658.2 XP_011536960.1 7 - 8 1096 115
XM_011538659.2 XP_011536961.1 10 - 11 1313 115
XM_017019800.1 XP_016875289.1 7 - 8 1533 115
XM_017019801.1 XP_016875290.1 6 - 7 660 115
XM_017019802.1 XP_016875291.1 5 - 6 593 115
XM_017019803.1 XP_016875292.1 6 - 7 660 115
XM_017019804.1 XP_016875293.1 5 - 6 593 115
GenBank mRNA AB030239.1 - 6 - 7 747 115
AB051853.1 - 8 - 9 1254 115
AK057573.1 - 5 - 6 867 115
AK092763.1 - 7 - 8 115
AK300910.1 - 5 - 6 860 115
AK301095.1 - 6 - 7 647 115
BC006107.1 - 11 - 12 1521 115
BC011820.1 - 6 - 7 115

View Full Product Details