Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARHGAP9
- Entrez Gene ID
-
64333
- Gene Name
- Rho GTPase activating protein 9
- Gene Aliases
-
10C, RGL1
- Chromosome Location
-
Chr.12: 57472255 - 57488814 on Build GRCh38
View Full Product Details