The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 5 - 6 | 908 | 191 |
XM_017016224.1 | XP_016871713.1 | 5 - 6 | 514 | 191 | |
GenBank mRNA | AK289413.1 | - | 5 - 6 | 854 | 191 |
AK307502.1 | - | 6 - 7 | 605 | 191 | |
BC010885.1 | - | 5 - 6 | 844 | 191 | |
BC065267.1 | - | 5 - 6 | 865 | 191 | |
BC080604.1 | - | 5 - 6 | 839 | 191 | |
BC111042.1 | - | 5 - 6 | 848 | 191 | |
BC126279.1 | - | 5 - 6 | 661 | 191 | |
BC126281.1 | - | 5 - 6 | 661 | 191 | |
FJ224324.1 | - | 5 - 6 | 751 | 191 | |
X65873.1 | - | 5 - 6 | 751 | 191 |