The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152718.2 | NP_689931.2 | 5 - 6 | 821 | 65 |
XM_006718460.1 | XP_006718523.1 | 5 - 6 | 821 | 65 | |
XM_006718462.1 | XP_006718525.1 | 5 - 6 | 821 | 65 | |
XM_011544817.1 | XP_011543119.1 | 5 - 6 | 821 | 65 | |
XM_017017341.1 | XP_016872830.1 | 5 - 6 | 821 | 65 | |
XM_017017342.1 | XP_016872831.1 | 4 - 5 | 414 | 65 | |
XM_017017343.1 | XP_016872832.1 | 5 - 6 | 546 | 65 | |
XM_017017344.1 | XP_016872833.1 | 5 - 6 | 821 | 65 | |
GenBank mRNA | AK056571.1 | - | 5 - 6 | 579 | 65 |
AK299583.1 | - | 4 - 5 | 728 | 65 | |
AY205235.1 | - | 3 - 4 | 326 | 65 | |
BC142624.1 | - | 5 - 6 | 568 | 65 |