The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005260302.3 | XP_005260359.1 | 3 - 4 | 629 | 123 |
XM_011528605.2 | XP_011526907.1 | 3 - 4 | 713 | 123 | |
XM_011528606.2 | XP_011526908.1 | 3 - 4 | 631 | 123 | |
XM_011528607.2 | XP_011526909.1 | 3 - 4 | 717 | 123 | |
XM_011528608.2 | XP_011526910.1 | 3 - 4 | 710 | 123 | |
XM_011528612.2 | XP_011526914.1 | 3 - 4 | 383 | 123 | |
XM_011528613.2 | XP_011526915.1 | 3 - 4 | 351 | 123 | |
XM_011528614.2 | XP_011526916.1 | 3 - 4 | 564 | 123 | |
GenBank mRNA | AK058202.1 | - | 3 - 4 | 388 | 123 |
AK298748.1 | - | 2 - 3 | 226 | 123 | |
BC057387.1 | - | 3 - 4 | 634 | 123 | |
BM547301.1 | - | 3 - 4 | 634 | 123 |