The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301860.1 | NP_001288789.1 | 4 - 5 | 716 | 74 |
NR_126051.1 | 4 - 5 | 716 | 74 | ||
XM_005260516.2 | XP_005260573.1 | 4 - 5 | 714 | 74 | |
XM_006723863.2 | XP_006723926.1 | 4 - 5 | 714 | 74 | |
XM_011528996.1 | XP_011527298.1 | 4 - 5 | 714 | 74 | |
XM_011528997.2 | XP_011527299.1 | 5 - 6 | 920 | 74 | |
XM_011528998.2 | XP_011527300.1 | 4 - 5 | 737 | 74 | |
XM_011528999.2 | XP_011527301.1 | 5 - 6 | 867 | 74 | |
XM_011529001.1 | XP_011527303.1 | 4 - 5 | 714 | 74 | |
XM_011529002.2 | XP_011527304.1 | 3 - 4 | 516 | 74 | |
XM_011529003.1 | XP_011527305.1 | 4 - 5 | 714 | 74 | |
XM_011529004.1 | XP_011527306.1 | 4 - 5 | 714 | 74 | |
XM_011529005.2 | XP_011527307.1 | 4 - 5 | 714 | 74 | |
XM_011529006.2 | XP_011527308.1 | 4 - 5 | 714 | 74 | |
GenBank mRNA | BC140806.1 | - | 4 - 5 | 597 | 74 |