Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
Chromosome Location
Chr.20: 41340779 - 41360585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301860.1 NP_001288789.1 9 - 10 1524 54
NR_126051.1 9 - 10 1524 54
XM_011528996.1 XP_011527298.1 9 - 10 1522 54
XM_011528997.2 XP_011527299.1 10 - 11 1728 54
XM_011528998.2 XP_011527300.1 9 - 10 1545 54
XM_011528999.2 XP_011527301.1 10 - 11 1675 54
XM_011529000.1 XP_011527302.1 9 - 10 1519 54
XM_011529001.1 XP_011527303.1 9 - 10 1522 54
XM_011529002.2 XP_011527304.1 8 - 9 1324 54
XM_011529003.1 XP_011527305.1 9 - 10 1522 54
XM_011529004.1 XP_011527306.1 9 - 10 1522 54
XM_011529005.2 XP_011527307.1 9 - 10 1522 54
XM_017028020.1 XP_016883509.1 9 - 10 1519 54
GenBank mRNA BC140806.1 - 9 - 10 1405 54

View Full Product Details