The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252675.1 | NP_001239604.1 | 11 - 12 | 1784 | 107 |
NM_001252676.1 | NP_001239605.1 | 10 - 11 | 1617 | 107 | |
NM_032501.3 | NP_115890.2 | 11 - 12 | 1790 | 107 | |
XM_006723659.1 | XP_006723722.1 | 7 - 8 | 1136 | 107 | |
XM_011529388.1 | XP_011527690.1 | 10 - 11 | 1575 | 107 | |
GenBank mRNA | AB058749.1 | - | 6 - 7 | 707 | 107 |
AK027817.1 | - | 5 - 6 | 613 | 107 | |
AK092295.1 | - | 9 - 10 | 1199 | 107 | |
AK125058.1 | - | 9 - 10 | 2993 | 107 | |
AK296306.1 | - | 10 - 11 | 1614 | 107 | |
AK314440.1 | - | 11 - 12 | 1713 | 107 | |
AL832939.1 | - | 7 - 8 | 859 | 107 | |
BC032076.1 | - | 4 - 5 | 478 | 107 | |
BC039261.1 | - | 11 - 12 | 1749 | 107 | |
BC044588.1 | - | 11 - 12 | 1709 | 107 | |
BC055008.1 | - | 11 - 12 | 1709 | 107 |