Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
Chromosome Location
Chr.20: 25006230 - 25058182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252675.1 NP_001239604.1 3 - 4 716 95
NM_001252676.1 NP_001239605.1 2 - 3 543 95
NM_001252677.1 NP_001239606.1 3 - 4 716 95
NM_032501.3 NP_115890.2 3 - 4 716 95
GenBank mRNA AK092295.1 - 1 - 2 133 95
AK125058.1 - 1 - 2 1919 95
AK127566.1 - 3 - 4 662 95
AK296306.1 - 2 - 3 540 95
AK314440.1 - 3 - 4 639 95
BC039261.1 - 3 - 4 675 95
BC044588.1 - 3 - 4 641 95
BC055008.1 - 3 - 4 641 95
BC114612.1 - 3 - 4 507 95

View Full Product Details