Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK2
Entrez Gene ID
5126
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
NEC 2, NEC-2, NEC2, PC2, SPC2
Chromosome Location
Chr.20: 17226107 - 17484578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201528.1 NP_001188457.1 2 - 3 422 74
NM_002594.4 NP_002585.2 1 - 2 527 74
GenBank mRNA AK312341.1 - 1 - 2 351 74
BC005815.2 - 1 - 2 428 74
BC040546.1 - 2 - 3 422 74
DA147868.1 - 1 - 2 527 74
DB173053.1 - 1 - 2 522 74
J05252.1 - 1 - 2 260 74

View Full Product Details