Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK2
Entrez Gene ID
5126
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
NEC 2, NEC-2, NEC2, PC2, SPC2
Chromosome Location
Chr.20: 17226107 - 17484578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201528.1 NP_001188457.1 9 - 10 1138 98
NM_001201529.2 NP_001188458.1 7 - 8 1138 98
NM_002594.4 NP_002585.2 8 - 9 1243 98
GenBank mRNA AK294200.1 - 7 - 8 1034 98
AK312341.1 - 8 - 9 1067 98
BC005815.2 - 8 - 9 1144 98
BC040546.1 - 9 - 10 1138 98
J05252.1 - 8 - 9 976 98

View Full Product Details