The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000059.3 | NP_000050.2 | 12 - 13 | 7165 | 172 |
GenBank mRNA | DQ897648.1 | - | 11 - 12 | 6938 | 172 |
U43746.1 | - | 12 - 13 | 7166 | 172 |