The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321193.1 | NP_001308122.1 | 13 - 14 | 1519 | 89 |
NM_001321194.1 | NP_001308123.1 | 14 - 15 | 1699 | 89 | |
NM_001321195.1 | NP_001308124.1 | 12 - 13 | 1416 | 89 | |
NM_024665.5 | NP_078941.2 | 13 - 14 | 1580 | 89 | |
XM_005247775.2 | XP_005247832.1 | 12 - 13 | 1535 | 89 | |
XM_006713746.1 | XP_006713809.1 | 14 - 15 | 1589 | 89 | |
XM_011513141.1 | XP_011511443.1 | 14 - 15 | 1523 | 89 | |
XM_011513142.2 | XP_011511444.1 | 13 - 14 | 1504 | 89 | |
XM_011513143.2 | XP_011511445.1 | 12 - 13 | 1430 | 89 | |
XM_017007185.1 | XP_016862674.1 | 13 - 14 | 1517 | 89 | |
GenBank mRNA | AF268193.1 | - | 13 - 14 | 1413 | 89 |
AF314544.1 | - | 12 - 13 | 1379 | 89 | |
AK022956.1 | - | 13 - 14 | 1479 | 89 | |
AK296946.1 | - | 11 - 12 | 1161 | 89 | |
BC113421.1 | - | 12 - 13 | 1322 | 89 | |
BX648935.1 | - | 13 - 14 | 1514 | 89 |