The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321193.1 | NP_001308122.1 | 6 - 7 | 830 | 78 |
NM_001321194.1 | NP_001308123.1 | 7 - 8 | 1010 | 78 | |
NM_001321195.1 | NP_001308124.1 | 5 - 6 | 727 | 78 | |
NM_024665.5 | NP_078941.2 | 6 - 7 | 891 | 78 | |
XM_005247775.2 | XP_005247832.1 | 5 - 6 | 846 | 78 | |
XM_006713746.1 | XP_006713809.1 | 7 - 8 | 900 | 78 | |
XM_011513141.1 | XP_011511443.1 | 7 - 8 | 834 | 78 | |
XM_011513142.2 | XP_011511444.1 | 6 - 7 | 815 | 78 | |
XM_011513143.2 | XP_011511445.1 | 5 - 6 | 741 | 78 | |
XM_017007185.1 | XP_016862674.1 | 6 - 7 | 828 | 78 | |
GenBank mRNA | AF268193.1 | - | 6 - 7 | 724 | 78 |
AF314544.1 | - | 5 - 6 | 690 | 78 | |
AK022956.1 | - | 6 - 7 | 790 | 78 | |
BC113421.1 | - | 5 - 6 | 633 | 78 | |
BX648935.1 | - | 6 - 7 | 825 | 78 |